Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105046.3 | NP_564815.1 | 1 - 2 | 629 | 64 | |
NM_202353.1 | NP_974082.1 | 1 - 2 | 263 | 64 | ||
GenBank mRNA | AY062807.1 | - | 1 - 2 | 216 | 64 | |
BT008895.1 | - | 1 - 2 | 151 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |