Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334122.1 | NP_001319310.1 | 4 - 5 | 1871 | 103 | |
NM_105047.2 | NP_176557.1 | 3 - 4 | 1421 | 103 | ||
GenBank mRNA | AY357947.1 | - | 3 - 4 | 1421 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |