Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334122.1 | NP_001319310.1 | 5 - 6 | 1941 | 100 | |
NM_105047.2 | NP_176557.1 | 4 - 5 | 1491 | 100 | ||
GenBank mRNA | AY357947.1 | - | 4 - 5 | 1491 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |