Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334122.1 | NP_001319310.1 | 7 - 8 | 2183 | 90 | |
NM_105047.2 | NP_176557.1 | 6 - 7 | 1733 | 90 | ||
GenBank mRNA | AY357947.1 | - | 6 - 7 | 1733 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |