Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHA-E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334149.1 | NP_001320763.1 | 1 - 2 | 163 | 126 | |
NM_001334150.1 | NP_001320764.1 | 1 - 2 | 180 | 126 | ||
NM_105094.3 | NP_176602.1 | 1 - 2 | 197 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |