Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHA-E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334149.1 | NP_001320763.1 | 2 - 3 | 271 | 69 | |
NM_001334150.1 | NP_001320764.1 | 2 - 3 | 288 | 69 | ||
NM_105094.3 | NP_176602.1 | 2 - 3 | 305 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |