Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105114.3 | NP_176621.1 | 7 - 8 | 1878 | 109 | |
GenBank mRNA | AF372940.1 | - | 7 - 8 | 1707 | 109 | |
AY143945.1 | - | 7 - 8 | 1573 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |