Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334162.1 | NP_001319319.1 | 8 - 9 | 719 | 67 | |
NM_001334163.1 | NP_001322653.1 | 8 - 9 | 695 | 67 | ||
NM_105123.2 | NP_176629.1 | 7 - 8 | 594 | 67 | ||
GenBank mRNA | AF290433.1 | - | 7 - 8 | 594 | 67 | |
AF411957.1 | - | 7 - 8 | 594 | 67 | ||
BT026348.1 | - | 7 - 8 | 594 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |