Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP89A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105169.3 | NP_176675.1 | 1 | 1902 | 99 | |
GenBank mRNA | AY035078.1 | - | 1 | 1847 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |