Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036158.2 | NP_001031235.1 | 2 - 3 | 518 | 113 | |
NM_105173.4 | NP_176679.2 | 1 - 2 | 358 | 113 | ||
GenBank mRNA | AY070446.1 | - | 1 - 2 | 319 | 113 | |
HM776216.1 | - | 1 - 2 | 206 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Arabidopsis thaliana Probe spans exons |