Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334199.1 | NP_001322349.1 | 4 - 5 | 813 | 115 | |
NM_105199.3 | NP_564846.1 | 4 - 5 | 658 | 115 | ||
GenBank mRNA | AY045845.1 | - | 4 - 5 | 426 | 115 | |
AY117153.1 | - | 4 - 5 | 417 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |