Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334199.1 | NP_001322349.1 | 5 - 6 | 909 | 93 | |
NM_105199.3 | NP_564846.1 | 5 - 6 | 754 | 93 | ||
GenBank mRNA | AY045845.1 | - | 5 - 6 | 522 | 93 | |
AY117153.1 | - | 5 - 6 | 513 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |