Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125073.2 | NP_001118545.1 | 3 - 4 | 913 | 130 | |
NM_001337304.1 | NP_001319436.1 | 3 - 4 | 868 | 130 | ||
NM_001337305.1 | NP_001319437.1 | 4 - 5 | 958 | 130 | ||
GenBank mRNA | KJ138849.1 | - | 3 - 4 | 415 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |