Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_110977.3 | NP_186761.1 | 1 - 2 | 841 | 90 | |
GenBank mRNA | AY094438.1 | - | 1 - 2 | 579 | 90 | |
BT002753.1 | - | 1 - 2 | 547 | 90 | ||
U83500.1 | - | 1 - 2 | 567 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |