Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337323.1 | NP_001319439.1 | 7 - 8 | 1133 | 96 | |
NM_001337324.1 | NP_001327637.1 | 6 - 7 | 1031 | 96 | ||
NM_001337325.1 | NP_001327636.1 | 7 - 8 | 1087 | 96 | ||
NM_110980.4 | NP_186764.1 | 6 - 7 | 1031 | 96 | ||
GenBank mRNA | AK316897.1 | - | 6 - 7 | 879 | 96 | |
AY093218.1 | - | 7 - 8 | 994 | 96 | ||
BT001239.1 | - | 5 - 6 | 420 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |