Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337323.1 | NP_001319439.1 | 8 - 9 | 1209 | 74 | |
NM_001337324.1 | NP_001327637.1 | 7 - 8 | 1107 | 74 | ||
NM_001337325.1 | NP_001327636.1 | 8 - 9 | 1163 | 74 | ||
NM_110980.4 | NP_186764.1 | 7 - 8 | 1107 | 74 | ||
GenBank mRNA | AK316897.1 | - | 7 - 8 | 955 | 74 | |
AY093218.1 | - | 8 - 9 | 1070 | 74 | ||
BT001239.1 | - | 6 - 7 | 496 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |