Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_110994.4 | NP_186777.1 | 2 - 3 | 319 | 84 | |
GenBank mRNA | AY037217.1 | - | 2 - 3 | 233 | 84 | |
AY113169.1 | - | 2 - 3 | 116 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |