Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202860.2 | NP_001189789.1 | 12 - 13 | 2123 | 70 | |
NM_110998.5 | NP_186781.4 | 12 - 13 | 2163 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |