Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036545.1 | NP_001031622.1 | 3 - 4 | 453 | 132 | |
NM_001125502.1 | NP_001118974.1 | 5 - 6 | 737 | 132 | ||
NM_001203783.2 | NP_001190712.1 | 5 - 6 | 774 | 132 | ||
NM_001340823.1 | NP_001329480.1 | 5 - 6 | 747 | 132 | ||
NM_001340824.1 | NP_001329479.1 | 5 - 6 | 747 | 132 | ||
NM_001340825.1 | NP_001329481.1 | 5 - 6 | 774 | 132 | ||
NM_117371.1 | NP_193038.1 | 3 - 4 | 453 | 132 | ||
NM_179039.4 | NP_849370.1 | 5 - 6 | 859 | 132 | ||
GenBank mRNA | AF360324.1 | - | 5 - 6 | 708 | 132 | |
AY056332.1 | - | 3 - 4 | 240 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |