Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117403.5 | NP_193066.4 | 1 - 2 | 129 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |