Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340854.1 | NP_001328592.1 | 11 - 12 | 1708 | 116 | |
NM_001340855.1 | NP_001328591.1 | 12 - 13 | 2125 | 116 | ||
NM_117408.4 | NP_567403.1 | 12 - 13 | 1697 | 116 | ||
NM_179042.1 | NP_849373.1 | 12 - 13 | 1636 | 116 | ||
GenBank mRNA | AY059783.1 | - | 12 - 13 | 1635 | 116 | |
AY117270.1 | - | 11 - 12 | 1136 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |