Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117409.5 | NP_193071.2 | 8 - 9 | 1595 | 87 | |
NM_179209.2 | NP_849540.1 | 8 - 9 | 1443 | 87 | ||
GenBank mRNA | AY062505.1 | - | 8 - 9 | 1443 | 87 | |
BT008379.1 | - | 7 - 8 | 1313 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |