Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117416.3 | NP_567404.1 | 8 - 9 | 1174 | 98 | |
GenBank mRNA | AF129478.1 | - | 8 - 9 | 1106 | 98 | |
BT010589.1 | - | 8 - 9 | 1097 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |