Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117479.4 | NP_193140.1 | 3 - 4 | 1065 | 113 | |
GenBank mRNA | AF370130.1 | - | 3 - 4 | 840 | 113 | |
AY040046.1 | - | 3 - 4 | 799 | 113 | ||
AY065104.1 | - | 3 - 4 | 834 | 113 | ||
AY065108.1 | - | 3 - 4 | 842 | 113 | ||
BT000385.1 | - | 3 - 4 | 799 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |