Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117488.4 | NP_193147.1 | 2 - 3 | 288 | 98 | |
GenBank mRNA | AF395067.1 | - | 2 - 3 | 167 | 98 | |
L32874.1 | - | 2 - 3 | 220 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |