Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340918.1 | NP_001328596.1 | 12 - 13 | 1725 | 116 | |
NM_117511.5 | NP_193170.1 | 13 - 14 | 1461 | 116 | ||
GenBank mRNA | AF250343.1 | - | 13 - 14 | 1285 | 116 | |
AY052364.1 | - | 13 - 14 | 1431 | 116 | ||
AY060528.1 | - | 13 - 14 | 1131 | 116 | ||
AY943850.1 | - | 13 - 14 | 1131 | 116 | ||
X78818.1 | - | 1302 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |