Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117550.5 | NP_193204.1 | 4 - 5 | 1594 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |