Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340965.1 | NP_001328646.1 | 2 - 3 | 841 | 101 | |
NM_001340966.1 | NP_001328647.1 | 2 - 3 | 830 | 101 | ||
NM_117560.3 | NP_193211.2 | 2 - 3 | 720 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |