Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSUV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340972.1 | NP_001328228.1 | 14 - 15 | 1858 | 103 | |
NM_117564.4 | NP_193215.2 | 14 - 15 | 1943 | 103 | ||
GenBank mRNA | AJ132843.1 | - | 14 - 15 | 1556 | 103 | |
AY139995.1 | - | 14 - 15 | 1858 | 103 | ||
BT008910.1 | - | 14 - 15 | 1535 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |