Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSUV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340972.1 | NP_001328228.1 | 1 - 2 | 380 | 72 | |
NM_117564.4 | NP_193215.2 | 1 - 2 | 465 | 72 | ||
GenBank mRNA | AJ132843.1 | - | 1 - 2 | 78 | 72 | |
AY139995.1 | - | 1 - 2 | 380 | 72 | ||
BT008910.1 | - | 1 - 2 | 57 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |