Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036570.1 | NP_001031647.1 | 16 - 17 | 2669 | 110 | |
NM_001084926.1 | NP_001078395.1 | 15 - 16 | 2606 | 110 | ||
NM_001084927.2 | NP_001078396.1 | 14 - 15 | 2658 | 110 | ||
NM_001125515.1 | NP_001118987.1 | 14 - 15 | 2551 | 110 | ||
NM_001341051.1 | NP_001328869.1 | 14 - 15 | 2777 | 110 | ||
NM_117643.4 | NP_193288.2 | 13 - 14 | 2407 | 110 | ||
NM_179060.4 | NP_849391.2 | 15 - 16 | 2645 | 110 | ||
GenBank mRNA | AK317187.1 | - | 15 - 16 | 2634 | 110 | |
AK317578.1 | - | 13 - 14 | 2325 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |