Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341063.1 | NP_001329006.1 | 9 - 10 | 1404 | 72 | |
NM_001341064.1 | NP_001329005.1 | 9 - 10 | 1160 | 72 | ||
NM_001341065.1 | NP_001329007.1 | 7 - 8 | 875 | 72 | ||
NM_117648.8 | NP_193292.3 | 10 - 11 | 1197 | 72 | ||
GenBank mRNA | EU915246.1 | - | 10 - 11 | 1197 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |