Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341063.1 | NP_001329006.1 | 16 - 17 | 2004 | 66 | |
NM_001341064.1 | NP_001329005.1 | 16 - 17 | 1760 | 66 | ||
NM_001341065.1 | NP_001329007.1 | 14 - 15 | 1475 | 66 | ||
NM_117648.8 | NP_193292.3 | 17 - 18 | 1797 | 66 | ||
GenBank mRNA | EU915246.1 | - | 17 - 18 | 1797 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |