Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203809.1 | NP_001190738.2 | 4 - 5 | 693 | 65 | |
NM_117667.9 | NP_193311.6 | 4 - 5 | 736 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |