Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117679.2 | NP_193322.2 | 1 - 2 | 263 | 77 | |
GenBank mRNA | Y11188.1 | - | 263 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |