Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117679.2 | NP_193322.2 | 5 - 6 | 1295 | 88 | |
GenBank mRNA | Y11188.1 | - | 1295 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |