Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117706.3 | NP_193348.1 | 14 - 15 | 1394 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |