Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341097.1 | NP_001319958.1 | 28 - 29 | 4445 | 77 | |
NM_117729.8 | NP_193367.7 | 27 - 28 | 4397 | 77 | ||
GenBank mRNA | AF465831.1 | - | 27 - 28 | 4330 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons |