Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341098.1 | NP_001328805.1 | 1 - 2 | 109 | 101 | |
NM_117730.3 | NP_567492.1 | 2 - 3 | 173 | 101 | ||
GenBank mRNA | AF192884.1 | - | 1 - 2 | 109 | 101 | |
AK176189.1 | - | 2 - 3 | 173 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |