Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036574.1 | NP_001031651.1 | 2 - 3 | 521 | 124 | |
NM_001341133.1 | NP_001329376.1 | 2 - 3 | 678 | 124 | ||
NM_001341134.1 | NP_001329377.1 | 2 - 3 | 678 | 124 | ||
NM_117775.4 | NP_567511.3 | 2 - 3 | 678 | 124 | ||
GenBank mRNA | BT030340.1 | - | 2 - 3 | 433 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |