Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117787.4 | NP_567517.1 | 12 - 13 | 1219 | 97 | |
NM_202834.2 | NP_974563.1 | 11 - 12 | 1321 | 97 | ||
GenBank mRNA | AY034902.1 | - | 12 - 13 | 1144 | 97 | |
AY063047.1 | - | 11 - 12 | 913 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |