Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGLYAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117809.6 | NP_193438.2 | 4 - 5 | 492 | 69 | |
GenBank mRNA | AK118019.1 | - | 4 - 5 | 377 | 69 | |
GQ303359.1 | - | 4 - 5 | 331 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |