Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP70-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117827.3 | NP_193455.2 | 4 - 5 | 556 | 109 | |
GenBank mRNA | AK117336.1 | - | 4 - 5 | 470 | 109 | |
AM086442.1 | - | 4 - 5 | 443 | 109 | ||
BT005901.1 | - | 4 - 5 | 385 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |