Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117878.3 | NP_567535.1 | 3 - 4 | 919 | 152 | |
GenBank mRNA | AY048690.1 | - | 3 - 4 | 698 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |