Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117899.3 | NP_567544.1 | 3 - 4 | 1001 | 84 | |
GenBank mRNA | AF302668.1 | - | 3 - 4 | 972 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |