Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP5PII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341251.1 | NP_001319979.1 | 8 - 9 | 1922 | 115 | |
NM_117911.4 | NP_567547.1 | 8 - 9 | 2013 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |