Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341257.1 | NP_001328203.1 | 2 - 3 | 696 | 92 | |
NM_117915.7 | NP_193539.6 | 2 - 3 | 277 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |