Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341273.1 | NP_001329801.1 | 10 - 11 | 2593 | 80 | |
NM_117939.5 | NP_193563.3 | 10 - 11 | 1969 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |