Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAVE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341297.1 | NP_001328468.1 | 2 - 3 | 520 | 109 | |
NM_001341298.1 | NP_001328469.1 | 2 - 3 | 500 | 109 | ||
NM_001341299.1 | NP_001328466.1 | 2 - 3 | 496 | 109 | ||
NM_001341300.1 | NP_001328467.1 | 2 - 3 | 500 | 109 | ||
NM_117976.4 | NP_193595.3 | 2 - 3 | 500 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |