Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343715.1 | NP_001330213.1 | 15 - 16 | 2110 | 77 | |
NM_147883.6 | NP_680188.1 | 15 - 16 | 2110 | 77 | ||
GenBank mRNA | BT014965.1 | - | 13 - 14 | 1908 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |